missing translation for 'onlineSavingsMsg'
Learn More

9-Decen-1-ol, 99%, Thermo Scientific™

Chemical Identifiers

CAS 13019-22-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
InChI Key QGFSQVPRCWJZQK-UHFFFAOYSA-N
Synonym 9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol
PubChem CID 25612
IUPAC Name dec-9-en-1-ol
SMILES C=CCCCCCCCCO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC152640050
SDS View Documents
Thermo Scientific Chemicals
152640050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC152640250
SDS View Documents
Thermo Scientific Chemicals
152640250
25g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC152641000
View Documents
Thermo Scientific Chemicals
152641000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13019-22-2
156.27
9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol
dec-9-en-1-ol
C10H20O
QGFSQVPRCWJZQK-UHFFFAOYSA-N
25612
C=CCCCCCCCCO

Specifications

13019-22-2
100.0
234.0°C to 238.0°C
Authentic
Glass bottle
1.4470 to 1.4490
5g
0.845
Solubility in water: insoluble. Other solubilities: soluble in alcohol
C=CCCCCCCCCO
156.27
11.4 mPa.s (20°C)
99%
98.5
0.8450g/mL
104°C
92% min. (main isomer) (GC)
C10H20O
H2C=CH(CH2)8OH
01,451
9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol
QGFSQVPRCWJZQK-UHFFFAOYSA-N
dec-9-en-1-ol
25612
156.27
9-Decen-1-ol, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 235-878-6

RTECSNumber : HE2095000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only