missing translation for 'onlineSavingsMsg'
Learn More

9-Decen-1-ol, 90+%

Catalog No. AAA1949022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1949022 100 g
AAA1949014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1949022 Supplier Thermo Scientific Chemicals Supplier No. A1949022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13019-22-2 | C10H20O | 156.269 g/mol

It finds it uses in a wide range of fragrances, especially in fine fragrances and soaps, in floral, rose petal or herbal fragrances. 9-Decen-1-ol is used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semi fluorinated isoprene) block copolymers with -CF2H-terminated side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it uses in a wide range of fragrances, especially in fine fragrances and soaps, in floral, rose petal or herbal fragrances. 9-Decen-1-ol is used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semi fluorinated isoprene) block copolymers with -CF2H-terminated side groups.

Solubility
Soluble in water at 20°C 0.16g/L. Soluble in alcohol, dipropylene glycol, paraffin oil.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 13019-22-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00002992
InChI Key QGFSQVPRCWJZQK-UHFFFAOYSA-N
Synonym 9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol
PubChem CID 25612
IUPAC Name dec-9-en-1-ol
SMILES C=CCCCCCCCCO

Specifications

Melting Point -13°C
Density 0.847
Boiling Point 235°C
Flash Point 104°C (219°F)
Odor Pleasant
Refractive Index 1.448
Quantity 100 g
Beilstein 1750928
Solubility Information Soluble in water at 20°C 0.16g/L. Soluble in alcohol,dipropylene glycol,paraffin oil.
Formula Weight 156.27
Percent Purity ≥90%
Chemical Name or Material 9-Decen-1-ol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-878-6
RTECSNumber HE2095000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only