missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Cyanophenanthrene 98.0+%, TCI America™
$177.08 - $609.54
Chemical Identifiers
| CAS | 2510-55-6 |
|---|---|
| Molecular Formula | C15H9N |
| Molecular Weight (g/mol) | 203.244 |
| MDL Number | MFCD00001173 |
| InChI Key | CWFIYYOQYVYBPW-UHFFFAOYSA-N |
| Synonym | 9-cyanophenanthrene, 9-phenanthrenecarbonitrile, 9-phenanthronitrile, 9-cyano-phenanthrene, 9-phenanthrenenitrile, acmc-209gh1, dsstox_cid_29112, dsstox_rid_83331, dsstox_gsid_49256 |
| PubChem CID | 17279 |
| IUPAC Name | phenanthrene-9-carbonitrile |
| SMILES | C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)C#N |
Chemical Identifiers
| 2510-55-6 | |
| 203.244 | |
| CWFIYYOQYVYBPW-UHFFFAOYSA-N | |
| 17279 | |
| C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)C#N |
| C15H9N | |
| MFCD00001173 | |
| 9-cyanophenanthrene, 9-phenanthrenecarbonitrile, 9-phenanthronitrile, 9-cyano-phenanthrene, 9-phenanthrenenitrile, acmc-209gh1, dsstox_cid_29112, dsstox_rid_83331, dsstox_gsid_49256 | |
| phenanthrene-9-carbonitrile |
Specifications
| 2510-55-6 | |
| White-Yellow | |
| C15H9N | |
| 1 g | |
| CWFIYYOQYVYBPW-UHFFFAOYSA-N | |
| phenanthrene-9-carbonitrile | |
| 17279 | |
| ≥98.0% (GC) | |
| 9-Cyanophenanthrene |
| 110°C | |
| 175°C | |
| MFCD00001173 | |
| 9-cyanophenanthrene, 9-phenanthrenecarbonitrile, 9-phenanthronitrile, 9-cyano-phenanthrene, 9-phenanthrenenitrile, acmc-209gh1, dsstox_cid_29112, dsstox_rid_83331, dsstox_gsid_49256 | |
| C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)C#N | |
| 203.244 | |
| 203.24 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : SF7199000
TSCA : No