accessibility menu, dialog, popup

UserName

CAS RN 2510-55-6

N

CAS RN 2510-55-6

IUPAC Name: phenanthrene-9-carbonitrile
Synonyms: phenanthrene-9-carbonitrile
Molecular Weight (g/mol): 203.24
Molecular Formula: C15H9N
InChi Key: CWFIYYOQYVYBPW-UHFFFAOYSA-N
SMILES: N#CC1=CC2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results