missing translation for 'onlineSavingsMsg'
Learn More

9-Cyanophenanthrene 98.0+%, TCI America™

Catalog No. C10715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10715G 5 g
C10711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10715G Supplier TCI America Supplier No. C10715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2510-55-6
Molecular Formula C15H9N
Molecular Weight (g/mol) 203.244
MDL Number MFCD00001173
InChI Key CWFIYYOQYVYBPW-UHFFFAOYSA-N
Synonym 9-cyanophenanthrene, 9-phenanthrenecarbonitrile, 9-phenanthronitrile, 9-cyano-phenanthrene, 9-phenanthrenenitrile, acmc-209gh1, dsstox_cid_29112, dsstox_rid_83331, dsstox_gsid_49256
PubChem CID 17279
IUPAC Name phenanthrene-9-carbonitrile
SMILES C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)C#N

Specifications

Melting Point 110°C
Color White-Yellow
Boiling Point 175°C
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Cyanophenanthrene
RTECSNumber SF7199000
TSCA No