missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9-cis-Retinoic Acid, >98%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0215872505
Specifications
9-cis-Retinoic Acid | |
5300-03-8 | |
C20H28O2 | |
5mg | |
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C | |
300.44 | |
CHEBI:50648 | |
Powder |
Cell Biology and Analysis | |
≥98% | |
9 z-retinoic acid, 9-cis retinoic acid, 9-cis-retinoic acid, 9-cis-tretinoin, alitretinoin, panretin, panretin gel, panretyn, panrexin, toctino | |
SHGAZHPCJJPHSC-YCNIQYBTSA-N | |
(2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid | |
449171 | |
300.44 |
Chemical Identifiers
5300-03-8 | |
300.44 | |
9 z-retinoic acid, 9-cis retinoic acid, 9-cis-retinoic acid, 9-cis-tretinoin, alitretinoin, panretin, panretin gel, panretyn, panrexin, toctino | |
CHEBI:50648 | |
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C |
C20H28O2 | |
SHGAZHPCJJPHSC-YCNIQYBTSA-N | |
449171 | |
(2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid |
Safety and Handling
Recommended Storage : Store at -20°C.