missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-cis-Retinoic Acid, >98%, MP Biomedicals™
$209.14 - $1878.12
Chemical Identifiers
| Molecular Formula | C20H28O2 |
|---|---|
| Molecular Weight (g/mol) | 300.44 |
| InChI Key | SHGAZHPCJJPHSC-YCNIQYBTSA-N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15872501
|
MP Biomedicals Inc
0215872501 |
1 mg |
Each for $209.14
|
|
|||||
|
ICN15872505
|
MP Biomedicals Inc
0215872505 |
5 mg |
Each for $862.78
|
|
|||||
|
ICN15872510
|
MP Biomedicals Inc
0215872510 |
10 mg |
Each for $1,878.12
|
|
|||||
Chemical Identifiers
| C20H28O2 | |
| 300.44 | |
| SHGAZHPCJJPHSC-YCNIQYBTSA-N |
Specifications
| 5300-03-8 | |
| C20H28O2 | |
| alitretinoin, 9-cis-retinoic acid, panretin, 9-cis-tretinoin, panrexin, panretyn, panretin gel, toctino, 9-cis retinoic acid, 9 z-retinoic acid | |
| CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C | |
| 300.44 | |
| CHEBI:50648 | |
| Powder |
| ≥98% | |
| 1 mg | |
| SHGAZHPCJJPHSC-YCNIQYBTSA-N | |
| (2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid | |
| 449171 | |
| 300.44 | |
| 9-cis-Retinoic Acid, Cell Biology and Analysis |
Safety and Handling
Recommended Storage : Store at -20°C.