missing translation for 'onlineSavingsMsg'
Learn More

9-cis-Retinoic Acid, >98%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215872501

 View more versions of this product

Catalog No. ICN15872501


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

9-cis-Retinoic Acid
5300-03-8
C20H28O2
alitretinoin, 9-cis-retinoic acid, panretin, 9-cis-tretinoin, panrexin, panretyn, panretin gel, toctino, 9-cis retinoic acid, 9 z-retinoic acid
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C
300.44
CHEBI:50648
Powder
Cell Biology and Analysis
≥98%
1 mg
SHGAZHPCJJPHSC-YCNIQYBTSA-N
(2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
449171
300.44

Chemical Identifiers

C20H28O2
300.44
SHGAZHPCJJPHSC-YCNIQYBTSA-N
CHEBI:50648
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C.

SDS
Documents

Documents

Promotions

Promotions