missing translation for 'onlineSavingsMsg'
Learn More

9-cis-Astaxanthin, ≤10% (all - trans-Astaxanthin, HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012628
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012628 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012628 Supplier Merck Emd Millipore Supplier No. 518811MG

9-cis-Astaxanthin is an isomer of astaxanthin (3,3 -dihydroxy-β,β-carotene-4,4 -dione), belonging to the xanthophyll family of oxygenated derivatives of carotenoids.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 113085-04-4
Linear Formula C40H52O4
Synonym (3 S,3′S, 9-cis)-3,3′-Dihydroxy-beta,beta-carotene-4,4′-dione
Type Analytical Standard
Quantity 1 mg
Formula Weight 596.84
Percent Purity ≤10% (all - trans-Astaxanthin, HPLC)
Grade Analytical Standard