missing translation for 'onlineSavingsMsg'
Learn More

9-Chloromethylanthracene 98.0+%, TCI America™

Catalog No. C10461G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10461G 1 g
C10465G 5 g
C104625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C10461G Supplier TCI America Supplier No. C10461G

Chemical Identifiers

CAS 24463-19-2
Molecular Formula C15H11Cl
Molecular Weight (g/mol) 226.703
MDL Number MFCD00001263
InChI Key PCVRSXXPGXRVEZ-UHFFFAOYSA-N
Synonym 9-chloromethyl anthracene, anthracene, 9-chloromethyl, 9-anthracenylmethyl chloride, ccris 7993, 9-chloromethyanthracene, pubchem14854, 9-chlormethyl-anthracen, acmc-209tez
PubChem CID 32385
IUPAC Name 9-(chloromethyl)anthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl

Specifications

Melting Point 139°C
Quantity 1 g
UN Number 3261
Formula Weight 226.70
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Chloromethylanthracene
RTECSNumber CA9600000
TSCA No