missing translation for 'onlineSavingsMsg'
Learn More

9-Chloromethylanthracene 97.0+%, TCI America™

Catalog No. p-7118276
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A55021G 1 g
A55025G 5 g
2 options

Catalog No. A55021G

Supplier: TCI America A55021G

Only null left

Chemical Identifiers

CAS 24463-19-2
Molecular Formula C15H11Cl
Molecular Weight (g/mol) 226.703
MDL Number MFCD00001263
InChI Key PCVRSXXPGXRVEZ-UHFFFAOYSA-N
Synonym 9-chloromethyl anthracene, anthracene, 9-chloromethyl, 9-anthracenylmethyl chloride, ccris 7993, 9-chloromethyanthracene, pubchem14854, 9-chlormethyl-anthracen, acmc-209tez
PubChem CID 32385
IUPAC Name 9-(chloromethyl)anthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl

Specifications

Melting Point 140°C
Color Yellow
Quantity 1 g
UN Number 3261
Formula Weight 226.70
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Chloromethylanthracene [for HPLC Labeling]
RTECSNumber CA9600000
TSCA No