missing translation for 'onlineSavingsMsg'
Learn More

9-(Chloromethyl)anthracene, 98+%

Catalog No. p-7021298
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1357906 5 g
AAA1357914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1357906 Supplier Thermo Scientific Chemicals Supplier No. A1357906
Only null left

CAS: 24463-19-2 | C15H11Cl | 226.703 g/mol

9-(Chloromethyl)anthracene is used as a blocking group reagent used for carboxylic acids, phenols, mercaptans and thiophenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-(Chloromethyl)anthracene is used as a blocking group reagent used for carboxylic acids, phenols, mercaptans and thiophenols.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 24463-19-2
Molecular Formula C15H11Cl
Molecular Weight (g/mol) 226.703
MDL Number MFCD00001263
InChI Key PCVRSXXPGXRVEZ-UHFFFAOYSA-N
Synonym 9-chloromethyl anthracene, anthracene, 9-chloromethyl, 9-anthracenylmethyl chloride, ccris 7993, 9-chloromethyanthracene, pubchem14854, 9-chlormethyl-anthracen, acmc-209tez
PubChem CID 32385
IUPAC Name 9-(chloromethyl)anthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl

Specifications

Melting Point 138°C to 142°C
Quantity 5 g
UN Number UN3261
Beilstein 1873394
Solubility Information Soluble in chloroform. Insoluble in water.
Formula Weight 226.71
Percent Purity ≥98%
Chemical Name or Material 9-(Chloromethyl)anthracene
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 246-274-7
RTECSNumber CA9600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only