missing translation for 'onlineSavingsMsg'
Learn More

9-Bromophenanthrene, 98%

CAS: 573-17-1 | C14H9Br | 257.13 g/mol

Supplier:  Thermo Scientific Chemicals L0903314

 View more versions of this product

Catalog No. AAL0903314

Only null left
Explore available promotions

Description

Description

9-Bromophenanthrene is used as halogenated building block. Isotope labelled 9-Bromophenanthrene (B687200), a halogenated polycyclic aromatic hydrocarbon that has been seen to have a room temperature phosphorescence that can be induced by β-cyclodextrin (β-CD) in the presence of cyclohexane; however, trace Fe(III) causes a decrease of the RTP emission.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Bromophenanthrene is used as halogenated building block. Isotope labelled 9-Bromophenanthrene (B687200), a halogenated polycyclic aromatic hydrocarbon that has been seen to have a room temperature phosphorescence that can be induced by β-cyclodextrin (β-CD) in the presence of cyclohexane; however, trace Fe(III) causes a decrease of the RTP emission.

Solubility
Soluble in toluene, and chloroform (50 mg/ml).

Notes
Incompatible with strong oxidizing agents. Store away from oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

9-Bromophenanthrene
62°C to 66°C
C14H9Br
25 g
9-phenanthryl bromide, phenanthrene, 9-bromo, 9-phenathryl bromide, 9-bromo-phenanthrene, 9-bromo phenanthrene, 9-bromophenathrene, 9-bromanylphenanthrene, pubchem13993, acmc-1b0zz, 9-bromophenanthrene
RSQXKVWKJVUZDG-UHFFFAOYSA-N
9-bromophenanthrene
11309
98%
573-17-1
190°C (1.2 mmHg)
MFCD00001174
1869927
Soluble in toluene,and chloroform (50mg/ml).
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)Br
257.13
257.14

Chemical Identifiers

573-17-1
257.13
RSQXKVWKJVUZDG-UHFFFAOYSA-N
11309
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)Br
C14H9Br
MFCD00001174
9-phenanthryl bromide, phenanthrene, 9-bromo, 9-phenathryl bromide, 9-bromo-phenanthrene, 9-bromo phenanthrene, 9-bromophenathrene, 9-bromanylphenanthrene, pubchem13993, acmc-1b0zz, 9-bromophenanthrene
9-bromophenanthrene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 209-351-6

RTECSNumber : SF7197000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only