Learn More
9-Bromophenanthrene, 98%
Description
9-Bromophenanthrene is used as halogenated building block. Isotope labelled 9-Bromophenanthrene (B687200), a halogenated polycyclic aromatic hydrocarbon that has been seen to have a room temperature phosphorescence that can be induced by β-cyclodextrin (β-CD) in the presence of cyclohexane; however, trace Fe(III) causes a decrease of the RTP emission.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
9-Bromophenanthrene is used as halogenated building block. Isotope labelled 9-Bromophenanthrene (B687200), a halogenated polycyclic aromatic hydrocarbon that has been seen to have a room temperature phosphorescence that can be induced by β-cyclodextrin (β-CD) in the presence of cyclohexane; however, trace Fe(III) causes a decrease of the RTP emission.
Solubility
Soluble in toluene, and chloroform (50 mg/ml).
Notes
Incompatible with strong oxidizing agents. Store away from oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Spécifications
Spécifications
| Melting Point | 62°C to 66°C |
| Boiling Point | 190°C (1.2 mmHg) |
| Quantity | 5 g |
| Beilstein | 1869927 |
| Solubility Information | Soluble in toluene,and chloroform (50mg/ml). |
| Formula Weight | 257.14 |
| Percent Purity | 98% |
| Chemical Name or Material | 9-Bromophenanthrene |
Sécurité et manipulation
| EINECSNumber | 209-351-6 |
| RTECSNumber | SF7197000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only