missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Bromofluorene 98.0+%, TCI America™
$78.29 - $225.56
Chemical Identifiers
| CAS | 1940-57-4 |
|---|---|
| Molecular Formula | C13H9Br |
| Molecular Weight (g/mol) | 245.119 |
| MDL Number | MFCD00001133 |
| InChI Key | AHCDKANCCBEQJJ-UHFFFAOYSA-N |
| Synonym | 9-bromofluorene, fluorene, 9-bromo, 9h-fluorene, 9-bromo, 9-fluorenyl bromide, monobromofluorene, 9-bromo-fluoren, 9-brom-9h-fluoren, acmc-1bqkc, 9-bromanyl-9h-fluorene |
| PubChem CID | 16024 |
| IUPAC Name | 9-bromo-9H-fluorene |
| SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)Br |
Chemical Identifiers
| 1940-57-4 | |
| 245.119 | |
| AHCDKANCCBEQJJ-UHFFFAOYSA-N | |
| 16024 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)Br |
| C13H9Br | |
| MFCD00001133 | |
| 9-bromofluorene, fluorene, 9-bromo, 9h-fluorene, 9-bromo, 9-fluorenyl bromide, monobromofluorene, 9-bromo-fluoren, 9-brom-9h-fluoren, acmc-1bqkc, 9-bromanyl-9h-fluorene | |
| 9-bromo-9H-fluorene |
Specifications
| 1940-57-4 | |
| Yellow | |
| MFCD00001133 | |
| 1759 | |
| AHCDKANCCBEQJJ-UHFFFAOYSA-N | |
| 9-bromo-9H-fluorene | |
| 16024 | |
| ≥98.0% (GC,T) | |
| 9-Bromofluorene |
| 104°C | |
| C13H9Br | |
| 5 g | |
| 9-bromofluorene, fluorene, 9-bromo, 9h-fluorene, 9-bromo, 9-fluorenyl bromide, monobromofluorene, 9-bromo-fluoren, 9-brom-9h-fluoren, acmc-1bqkc, 9-bromanyl-9h-fluorene | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)Br | |
| 245.119 | |
| 245.12 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : LL5890000
TSCA : No