missing translation for 'onlineSavingsMsg'
Learn More

9-Bromoanthracene 95.0+%, TCI America™

Catalog No. B08725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08725G 5 g
B087225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08725G Supplier TCI America Supplier No. B08725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1564-64-3
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD00001243
InChI Key ZIRVQSRSPDUEOJ-UHFFFAOYSA-N
Synonym anthracene, 9-bromo, 9-bromo anthracene, 9-bromo-anthracene, 9-anthracenyl bromide, 9-bromanthracen, pubchem9183, 9-bromoanthrene, acmc-209ddx, 9-bromoanthracene, dsstox_cid_29080
PubChem CID 74062
IUPAC Name 9-bromoanthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Br

Specifications

Melting Point 101°C
Quantity 5 g
Formula Weight 257.13
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromoanthracene