missing translation for 'onlineSavingsMsg'
Learn More

9-Bromoanthracene, 96%

Catalog No. AC106640050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106640050 5 g Glass bottle
AC106640250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106640050 Supplier Thermo Scientific Chemicals Supplier No. 106640050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1564-64-3 | C14H9Br | 257.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1564-64-3
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD00001243
InChI Key ZIRVQSRSPDUEOJ-UHFFFAOYSA-N
Synonym anthracene, 9-bromo, 9-bromo anthracene, 9-bromo-anthracene, 9-anthracenyl bromide, 9-bromanthracen, pubchem9183, 9-bromoanthrene, acmc-209ddx, 9-bromoanthracene, dsstox_cid_29080
PubChem CID 74062
IUPAC Name 9-bromoanthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 98°C to 100°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 05, 665
Solubility Information Solubility in water: insoluble
Formula Weight 257.13
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 9-Bromoanthracene, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-359-3

RUO – Research Use Only