missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-9,10-dihydro-9,10-ethenoanthracene 98.0+%, TCI America™

Catalog No. B42391G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42391G 1 g
B42395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42391G Supplier TCI America Supplier No. B42391G
Only null left

Chemical Identifiers

CAS 126690-96-8
Molecular Formula C16H11Br
Molecular Weight (g/mol) 283.168
InChI Key JNPBTNVWRDISGS-UHFFFAOYSA-N
Synonym 9-Bromodibenzobarrelene
PubChem CID 91972153
SMILES C1=CC=C2C(=C1)C3C=CC2(C4=CC=CC=C34)Br

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 1 g
Formula Weight 283.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-9,10-dihydro-9,10-ethenoanthracene