missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-7,7-dimethyl-7H-benzo[c]fluorene 98.0+%, TCI America™

Catalog No. b4952200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4952200MG 200 mg
B49521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4952200MG Supplier TCI America Supplier No. B4952200MG
Only null left

Chemical Identifiers

CAS 1198396-46-1
Molecular Formula C19H15Br
Molecular Weight (g/mol) 323.23
MDL Number MFCD27980277
InChI Key SNUKBTHMZLUJKR-UHFFFAOYSA-N
PubChem CID 77231394
IUPAC Name 9-bromo-7,7-dimethyl-7H-benzo[c]fluorene
SMILES CC1(C)C2=CC=C3C=CC=CC3=C2C2=C1C=C(Br)C=C2

Specifications

Melting Point 107°C
Color White
Quantity 200 mg
Formula Weight 323.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-7,7-dimethyl-7H-benzo[c]fluorene