missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Bromo-7,7-dimethyl-7H-benzo[c]fluorene 98.0+%, TCI America™
Supplier: TCI America B49521G
Specifications
| 9-Bromo-7,7-dimethyl-7H-benzo[c]fluorene | |
| 107°C | |
| C19H15Br | |
| 1 g | |
| CC1(C)C2=CC=C3C=CC=CC3=C2C2=C1C=C(Br)C=C2 | |
| 323.23 | |
| 323.23 | |
| Crystalline Powder |
| 1198396-46-1 | |
| White | |
| MFCD27980277 | |
| SNUKBTHMZLUJKR-UHFFFAOYSA-N | |
| 9-bromo-7,7-dimethyl-7H-benzo[c]fluorene | |
| 77231394 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1198396-46-1 | |
| 323.23 | |
| SNUKBTHMZLUJKR-UHFFFAOYSA-N | |
| 9-bromo-7,7-dimethyl-7H-benzo[c]fluorene |
| C19H15Br | |
| MFCD27980277 | |
| 77231394 | |
| CC1(C)C2=CC=C3C=CC=CC3=C2C2=C1C=C(Br)C=C2 |
Safety and Handling
TSCA : No