missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-10-[4-(1-naphthyl)phenyl]anthracene 98.0+%, TCI America™

Catalog No. B4570200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4570200MG 200 mg
B45701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4570200MG Supplier TCI America Supplier No. B4570200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1092390-01-6
Molecular Formula C30H19Br
Molecular Weight (g/mol) 459.386
MDL Number MFCD20486477
InChI Key AEIPFESDNDUXNK-UHFFFAOYSA-N
PubChem CID 59861528
IUPAC Name 9-bromo-10-(4-naphthalen-1-ylphenyl)anthracene
SMILES C1=CC=C2C(=C1)C=CC=C2C3=CC=C(C=C3)C4=C5C=CC=CC5=C(C6=CC=CC=C64)Br

Specifications

Melting Point 213°C
Quantity 200 mg
Formula Weight 459.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-10-[4-(1-naphthyl)phenyl]anthracene