accessibility menu, dialog, popup

UserName

CAS RN 1092390-01-6

IUPAC Name: 9-bromo-10-[4-(naphthalen-1-yl)phenyl]anthracene
Synonyms: 9-bromo-10-[4-(naphthalen-1-yl)phenyl]anthracene
Molecular Weight (g/mol): 459.39
Molecular Formula: C30H19Br
InChi Key: AEIPFESDNDUXNK-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=C(C2=CC=C(C=C2)C2=CC=CC3=CC=CC=C23)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results