missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-1-nonene 95.0+%, TCI America™

Catalog No. B32561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32561G 1 g
B32565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32561G Supplier TCI America Supplier No. B32561G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89359-54-6
Molecular Formula C9H17Br
Molecular Weight (g/mol) 205.14
MDL Number MFCD09037826
InChI Key RQXPBVHYVAOUBY-UHFFFAOYSA-N
Synonym 9-bromo-1-nonene, 1-nonene, 9-bromo, 8-nonenyl bromide, 1-bromo-8-nonene, 9-bromanylnon-1-ene
PubChem CID 11019998
IUPAC Name 9-bromonon-1-ene
SMILES BrCCCCCCCC=C

Specifications

Color Colorless
Boiling Point 101°C
Quantity 1 g
Formula Weight 205.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 9-Bromo-1-nonene
EINECSNumber (2)-0067
TSCA No
Recommended Storage Refrigerator