missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-1-nonene, 97%

Catalog No. p-7056050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6479203 1 g
AAH6479206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6479203 Supplier Thermo Scientific Chemicals Supplier No. H6479203

CAS: 89359-54-6 | C9H17Br | 205.14 g/mol

It's employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 89359-54-6
Molecular Formula C9H17Br
Molecular Weight (g/mol) 205.14
MDL Number MFCD09037826
InChI Key RQXPBVHYVAOUBY-UHFFFAOYSA-N
Synonym 9-bromo-1-nonene, 1-nonene, 9-bromo, 8-nonenyl bromide, 1-bromo-8-nonene, 9-bromanylnon-1-ene
PubChem CID 11019998
IUPAC Name 9-bromonon-1-ene
SMILES BrCCCCCCCC=C

Specifications

Color Colorless to Yellow
Boiling Point 100°C to 101°C (15 mmHg)
Linear Formula Br(CH2)7CH=CH2
Refractive Index 1.4675
Quantity 1 g
Beilstein 1746338
Formula Weight 205.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 9-Bromo-1-nonene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only