missing translation for 'onlineSavingsMsg'
Learn More

9-beta-D-Arabinofuranosyladenine, 99%

Catalog No. p-7043015
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6507606 5 g
AAJ6507614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6507606 Supplier Thermo Scientific Chemicals Supplier No. J6507606
Only null left

CAS: 5536-17-4 | C10H13N5O4 | 267.25 g/mol

Vidarabine is a neurotransmitter that acts as the preferred endogenous agonist at all adenosine receptor subtypes. Vidarabine is an inhibitor of A cyclase and DNA Polymerase. Used to study the roles of AMP-activated protein kinase (AMPK) in cell signaling. Cell-permeable adenylate cyclase inhibitor; in detergent-dispersed rat brain preparation, IC50 = 30 ?M. Clinically significant antiviral agent, especially against herpes simplex (HSV),1 by inhibition of DNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vidarabine is a neurotransmitter that acts as the preferred endogenous agonist at all adenosine receptor subtypes. Vidarabine is an inhibitor of A cyclase and DNA Polymerase. Used to study the roles of AMP-activated protein kinase (AMPK) in cell signaling. Cell-permeable adenylate cyclase inhibitor; in detergent-dispersed rat brain preparation, IC50 = 30 μM. Clinically significant antiviral agent, especially against herpes simplex (HSV),1 by inhibition of DNA polymerase.

Solubility
Soluble in DMF (10 mg/ml), 0.5 M HCl (50 mg/ml), DMSO (53 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), and water (3 mg/ml at 25°C).

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 5536-17-4
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00065471,MFCD00065471,MFCD00475667,MFCD00005752,MFCD00065471
InChI Key OIRDTQYFTABQOQ-UHFFFAOYNA-N
Synonym 3s,4r,5r-2-6-aminopurin-9-yl-5-hydroxymethyl oxolane-3,4-diol, prestwick_983, 9-lyxofuranosyladenine
PubChem CID 6420052
IUPAC Name (3S,4R,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN(C3OC(CO)C(O)C3O)C2=NC=N1

Specifications

Color White
Quantity 5 g
Beilstein 624881
Merck Index 14,9976
Solubility Information Soluble in DMF (10mg/ml),0.5 M HCl (50mg/ml),DMSO (53mg/ml at 25°C),ethanol (<1mg/ml at 25°C),and water (3mg/ml at 25°C).
Formula Weight 267.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 9-beta-D-Arabinofuranosyladenine
Hazard Category H361
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 226-893-9
RTECSNumber AU6200000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only