missing translation for 'onlineSavingsMsg'
Learn More

9-Benzylcarbazole-3-carboxaldehyde 97.0+%, TCI America™

Catalog No. B28045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28045G 5 g
B28041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28045G Supplier TCI America Supplier No. B28045G
Only null left

Chemical Identifiers

CAS 54117-37-2
Molecular Formula C20H15NO
Molecular Weight (g/mol) 285.35
MDL Number MFCD00403522
InChI Key GSNXZYWQXATWRX-UHFFFAOYSA-N
Synonym 9-Benzyl-3-formylcarbazole
PubChem CID 3091534
IUPAC Name 9-benzyl-9H-carbazole-3-carbaldehyde
SMILES O=CC1=CC2=C(C=C1)N(CC1=CC=CC=C1)C1=C2C=CC=C1

Specifications

Melting Point 134°C
Quantity 5 g
Formula Weight 285.35
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzylcarbazole-3-carboxaldehyde