accessibility menu, dialog, popup

UserName

NO

CAS RN 54117-37-2

IUPAC Name: 9-benzyl-9H-carbazole-3-carbaldehyde
Synonyms: 9-benzylcarbazole-3-carbaldehyde
Molecular Weight (g/mol): 285.35
Molecular Formula: C20H15NO
InChi Key: GSNXZYWQXATWRX-UHFFFAOYSA-N
SMILES: O=CC1=CC2=C(C=C1)N(CC1=CC=CC=C1)C1=C2C=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results