missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Benzoylcarbazole 98.0+%, TCI America™
$55.92 - $274.02
Chemical Identifiers
| CAS | 19264-68-7 |
|---|---|
| Molecular Formula | C19H13NO |
| Molecular Weight (g/mol) | 271.32 |
| MDL Number | MFCD00225468 |
| InChI Key | BUCKMWPLVBYQCQ-UHFFFAOYSA-N |
| PubChem CID | 95052 |
| IUPAC Name | 9-benzoyl-9H-carbazole |
| SMILES | O=C(N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=CC=C1 |
Chemical Identifiers
| 19264-68-7 | |
| 271.32 | |
| BUCKMWPLVBYQCQ-UHFFFAOYSA-N | |
| 9-benzoyl-9H-carbazole |
| C19H13NO | |
| MFCD00225468 | |
| 95052 | |
| O=C(N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=CC=C1 |
Specifications
| 19264-68-7 | |
| White-Yellow | |
| MFCD00225468 | |
| BUCKMWPLVBYQCQ-UHFFFAOYSA-N | |
| 9-benzoyl-9H-carbazole | |
| 95052 | |
| ≥98.0% (GC) | |
| 9-Benzoylcarbazole |
| 101°C | |
| C19H13NO | |
| 1 g | |
| O=C(N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=CC=C1 | |
| 271.32 | |
| 271.32 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : FE5780000
TSCA : No