accessibility menu, dialog, popup

UserName

CAS RN 19264-68-7

ON

CAS RN 19264-68-7

IUPAC Name: 9-benzoyl-9H-carbazole
Synonyms: 9-benzoylcarbazole
Molecular Weight (g/mol): 271.32
Molecular Formula: C19H13NO
InChi Key: BUCKMWPLVBYQCQ-UHFFFAOYSA-N
SMILES: O=C(N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results