missing translation for 'onlineSavingsMsg'
Learn More

9-Benzoylcarbazole 98.0+%, TCI America™

Catalog No. B28645G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28645G 5 g
B28641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28645G Supplier TCI America Supplier No. B28645G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19264-68-7
Molecular Formula C19H13NO
Molecular Weight (g/mol) 271.32
MDL Number MFCD00225468
InChI Key BUCKMWPLVBYQCQ-UHFFFAOYSA-N
PubChem CID 95052
IUPAC Name 9-benzoyl-9H-carbazole
SMILES O=C(N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=CC=C1

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
Formula Weight 271.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzoylcarbazole
RTECSNumber FE5780000
TSCA No