missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Benzoylcarbazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 101°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 271.32 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 9-Benzoylcarbazole |
Safety and Handling
| RTECSNumber | FE5780000 |
| TSCA | No |