missing translation for 'onlineSavingsMsg'
Learn More

9-Azajulolidine 97.0+%, TCI America™

Catalog No. A18541G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A18541G Supplier TCI America Supplier No. A18541G
Only null left

Chemical Identifiers

CAS 6052-72-8
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.247
InChI Key VARHFUNXFXTFII-UHFFFAOYSA-N
Synonym 2,3,6,7-Tetrahydro-1H,5H-9-azabenzo[ij]quinolizine
PubChem CID 11378792
SMILES C1CC2=CN=CC3=C2N(C1)CCC3

Specifications

Melting Point 72°C
Color White
Boiling Point 145°C
Quantity 1 g
Formula Weight 174.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Azajulolidine
TSCA No
Recommended Storage Refrigerator