accessibility menu, dialog, popup

UserName

CAS RN 6052-72-8

NN

CAS RN 6052-72-8

IUPAC Name: 1,7-diazatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene
Synonyms: 1,7-diazatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene
Molecular Weight (g/mol): 174.25
Molecular Formula: C11H14N2
InChi Key: VARHFUNXFXTFII-UHFFFAOYSA-N
SMILES: C1CN2CCCC3=C2C(C1)=CN=C3
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results