Learn More
9-Anthraldehyde, 98%
CAS: 642-31-9 | C15H10O | 206.24 g/mol
Supplier: Thermo Scientific Chemicals A1125822
Description
9-Anthraldehyde is used to prepare asymmetrical tridentate Schiff base ligands, 2-(9-anthrylmethyl-ideneamino)-4-methyl-phenol and functionalized ligand like 2-(anthracen-9-ylidene)-4,5-bis(diphenylphosphino)-4-cyclopentene-1,3-dione through Knoevenangel condensation with the diphosphine ligand 4,5-bis(diphenylphosphino)-4-cyclopentene-1,3-dione. It is also used as an intermediate for dyes, pigment, active pharmaceutical ingredients and other organic compounds. Further, it is involved in the Diels Alder reaction with benzenediazonium-2-carboxylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9-Anthraldehyde is used to prepare asymmetrical tridentate Schiff base ligands, 2-(9-anthrylmethyl-ideneamino)-4-methyl-phenol and functionalized ligand like 2-(anthracen-9-ylidene)-4,5-bis(diphenylphosphino)-4-cyclopentene-1,3-dione through Knoevenangel condensation with the diphosphine ligand 4,5-bis(diphenylphosphino)-4-cyclopentene-1,3-dione. It is also used as an intermediate for dyes, pigment, active pharmaceutical ingredients and other organic compounds. Further, it is involved in the Diels Alder reaction with benzenediazonium-2-carboxylate.
Solubility
Soluble in toluene.
Notes
Air sensitive. Incompatible with strong bases and strong oxidizing agents.
Specifications
| 9-Anthraldehyde | |
| 102°C to 106°C | |
| MFCD00001254 | |
| 639167 | |
| 9-anthraldehyde, 9-anthracenecarboxaldehyde, 9-anthrylaldehyde, 9-formylanthracene, 9-anthracenecarbaldehyde, 9-anthrylcarboxaldehyde, ccris 3165, nsc 15, anthracene-9-aldehyde, anthracene-9-carboxaldehyde | |
| YMNKUHIVVMFOFO-UHFFFAOYSA-N | |
| anthracene-9-carbaldehyde | |
| 69504 | |
| 98% |
| 642-31-9 | |
| C15H10O | |
| 100 g | |
| Air Sensitive | |
| Soluble in toluene. | |
| O=CC1=C2C=CC=CC2=CC2=CC=CC=C12 | |
| 206.24 | |
| 206.24 |
Chemical Identifiers
| 642-31-9 | |
| 206.24 | |
| YMNKUHIVVMFOFO-UHFFFAOYSA-N | |
| 69504 | |
| O=CC1=C2C=CC=CC2=CC2=CC=CC=C12 |
| C15H10O | |
| MFCD00001254 | |
| 9-anthraldehyde, 9-anthracenecarboxaldehyde, 9-anthrylaldehyde, 9-formylanthracene, 9-anthracenecarbaldehyde, 9-anthrylcarboxaldehyde, ccris 3165, nsc 15, anthracene-9-aldehyde, anthracene-9-carboxaldehyde | |
| anthracene-9-carbaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 211-383-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only