missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Anthracenecarboxylic Acid 97.0+%, TCI America™
$61.61 - $240.64
Chemical Identifiers
| CAS | 723-62-6 |
|---|---|
| Molecular Formula | C15H10O2 |
| Molecular Weight (g/mol) | 222.24 |
| MDL Number | MFCD00001257 |
| InChI Key | XGWFJBFNAQHLEF-UHFFFAOYSA-N |
| Synonym | 9-anthracenecarboxylic acid, 9-anthroic acid, anca, 9-carboxyanthracene, 9-ac, anthracene-10-carboxylic acid, 9-anthracene carboxylic acid, a9c, spectrum_001457, tocris-0963 |
| PubChem CID | 2201 |
| ChEBI | CHEBI:34507 |
| IUPAC Name | anthracene-9-carboxylic acid |
| SMILES | OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12 |
Chemical Identifiers
| 723-62-6 | |
| 222.24 | |
| XGWFJBFNAQHLEF-UHFFFAOYSA-N | |
| 2201 | |
| anthracene-9-carboxylic acid |
| C15H10O2 | |
| MFCD00001257 | |
| 9-anthracenecarboxylic acid, 9-anthroic acid, anca, 9-carboxyanthracene, 9-ac, anthracene-10-carboxylic acid, 9-anthracene carboxylic acid, a9c, spectrum_001457, tocris-0963 | |
| CHEBI:34507 | |
| OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12 |
Specifications
| 723-62-6 | |
| C15H10O2 | |
| 5 g | |
| XGWFJBFNAQHLEF-UHFFFAOYSA-N | |
| anthracene-9-carboxylic acid | |
| 2201 | |
| 222.24 | |
| Crystalline Powder |
| 219°C | |
| MFCD00001257 | |
| 9-anthracenecarboxylic acid, 9-anthroic acid, anca, 9-carboxyanthracene, 9-ac, anthracene-10-carboxylic acid, 9-anthracene carboxylic acid, a9c, spectrum_001457, tocris-0963 | |
| OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12 | |
| 222.24 | |
| CHEBI:34507 | |
| ≥97.0% (GC,T) | |
| 9-Anthracenecarboxylic Acid |
Safety and Handling
EINECSNumber : (4)-1876
RTECSNumber : CB8764000
TSCA : Yes