missing translation for 'onlineSavingsMsg'
Learn More

9-Anthracenecarboxylic Acid 97.0+%, TCI America™

Catalog No. A06905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06905G 5 g
A069025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06905G Supplier TCI America Supplier No. A06905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 723-62-6
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.24
MDL Number MFCD00001257
InChI Key XGWFJBFNAQHLEF-UHFFFAOYSA-N
Synonym 9-anthracenecarboxylic acid, 9-anthroic acid, anca, 9-carboxyanthracene, 9-ac, anthracene-10-carboxylic acid, 9-anthracene carboxylic acid, a9c, spectrum_001457, tocris-0963
PubChem CID 2201
ChEBI CHEBI:34507
IUPAC Name anthracene-9-carboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 219°C
Quantity 5 g
Formula Weight 222.24
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Anthracenecarboxylic Acid
EINECSNumber (4)-1876
RTECSNumber CB8764000
TSCA Yes