missing translation for 'onlineSavingsMsg'
Learn More

9-Anthracenecarboxaldehyde 99.0+%, TCI America™

Catalog No. p-7117053
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A077925G 25 g
A0779100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A077925G Supplier TCI America Supplier No. A077925G
Only null left

Chemical Identifiers

CAS 642-31-9
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001254
InChI Key YMNKUHIVVMFOFO-UHFFFAOYSA-N
Synonym 9-anthraldehyde, 9-anthracenecarboxaldehyde, 9-anthrylaldehyde, 9-formylanthracene, 9-anthracenecarbaldehyde, 9-anthrylcarboxaldehyde, ccris 3165, nsc 15, anthracene-9-aldehyde, anthracene-9-carboxaldehyde
PubChem CID 69504
IUPAC Name anthracene-9-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 107°C
Color Yellow
Quantity 25 g
Formula Weight 206.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Anthracenecarboxaldehyde