missing translation for 'onlineSavingsMsg'
Learn More

9-Anthracenecarboxaldehyde, 99%

CAS: 642-31-9 | C15H10O | 206.24 g/mol

$121.73 - $121.73

Chemical Identifiers

CAS 642-31-9
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001254
InChI Key YMNKUHIVVMFOFO-UHFFFAOYSA-N
Synonym 9-anthraldehyde, 9-anthracenecarboxaldehyde, 9-anthrylaldehyde, 9-formylanthracene, 9-anthracenecarbaldehyde, 9-anthrylcarboxaldehyde, ccris 3165, nsc 15, anthracene-9-aldehyde, anthracene-9-carboxaldehyde
PubChem CID 69504
IUPAC Name anthracene-9-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=CC2=CC=CC=C12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC153930250
SDS View Documents
Thermo Scientific Chemicals
153930250
25 g Glass bottle
Each for $121.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

642-31-9
206.24
YMNKUHIVVMFOFO-UHFFFAOYSA-N
69504
O=CC1=C2C=CC=CC2=CC2=CC=CC=C12
C15H10O
MFCD00001254
9-anthraldehyde, 9-anthracenecarboxaldehyde, 9-anthrylaldehyde, 9-formylanthracene, 9-anthracenecarbaldehyde, 9-anthrylcarboxaldehyde, ccris 3165, nsc 15, anthracene-9-aldehyde, anthracene-9-carboxaldehyde
anthracene-9-carbaldehyde

Specifications

642-31-9
Yellow
98.5% min. (HPLC)
C15H10O
25 g
Solubility in water: .. Other solubilities: soluble in chloroform
O=CC1=C2C=CC=CC2=CC2=CC=CC=C12
206.24
206.24
Crystalline Powder or Needles
103.0°C to 107.0°C
Authentic
Glass bottle
MFCD00001254
9-anthraldehyde, 9-anthracenecarboxaldehyde, 9-anthrylaldehyde, 9-formylanthracene, 9-anthracenecarbaldehyde, 9-anthrylcarboxaldehyde, ccris 3165, nsc 15, anthracene-9-aldehyde, anthracene-9-carboxaldehyde
YMNKUHIVVMFOFO-UHFFFAOYSA-N
anthracene-9-carbaldehyde
69504
99%
9-Anthracenecarboxaldehyde
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-383-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only