missing translation for 'onlineSavingsMsg'
Learn More

9-Anthraceneboronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™

Catalog No. A23285G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23285G 5 g
A23281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23285G Supplier TCI America Supplier No. A23285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100622-34-2
Molecular Formula C14H11BO2
Molecular Weight (g/mol) 222.05
MDL Number MFCD03425925
InChI Key VHHDLIWHHXBLBK-UHFFFAOYSA-N
Synonym 9-anthraceneboronic acid, anthracene-9-boronic acid, 9-anthracenylboronic acid, 9-anthrylboronic acid, 9-anthracene boronic acid, boronic acid, 9-anthracenyl, 9-boronoanthracene, 9-borono-anthracene, zlchem 414, pubchem7757
PubChem CID 15510213
IUPAC Name (anthracen-9-yl)boronic acid
SMILES OB(O)C1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 217°C
Quantity 5 g
Formula Weight 222.05
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Anthraceneboronic Acid (contains varying amounts of Anhydride)