missing translation for 'onlineSavingsMsg'
Learn More

9-Aminophenanthrene, 96%

Catalog No. p-7052782
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55773MD 250 mg
AAH5577303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH55773MD Supplier Thermo Scientific Chemicals Supplier No. H55773MD
Only null left

CAS: 947-73-9 | C14H11N | 193.249 g/mol

9-Aminophenanthrene can be used as a model ligand to study the binding of 9-aminophenanthrene to individual subsites within the active site of CYP(eryF). It is also used as a fluorescent-labeling reagent for free fatty acids (FFA),examination through HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Aminophenanthrene can be used as a model ligand to study the binding of 9-aminophenanthrene to individual subsites within the active site of CYP(eryF). It is also used as a fluorescent-labeling reagent for free fatty acids (FFA),examination through HPLC.

Solubility
It is slightly soluble in water 0.00581 mg/mL.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 947-73-9
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00001177
InChI Key KIHQWOBUUIPWAN-UHFFFAOYSA-N
Synonym 9-aminophenanthrene, 9-phenanthrenamine, 9-phenanthrylamine, phenanthren-9-ylamine, unii-7m9m5km2xy, ccris 7006, 7m9m5km2xy, 9ap, acmc-20alvr, phenanthren-9-yl-amine
PubChem CID 13695
ChEBI CHEBI:50475
IUPAC Name phenanthren-9-amine
SMILES C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)N

Specifications

Melting Point 137°C to 139°C
Color White to Brown
Quantity 250 mg
Solubility Information It is slightly soluble in water 0.00581mg/mL.
Formula Weight 193.24
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 9-Aminophenanthrene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-431-6
RTECSNumber SG0175000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only