accessibility menu, dialog, popup

UserName

H2N

CAS RN 947-73-9

IUPAC Name: phenanthren-9-amine
Synonyms: 9-aminophenanthrene
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: KIHQWOBUUIPWAN-UHFFFAOYSA-N
SMILES: NC1=CC2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results