missing translation for 'onlineSavingsMsg'
Learn More

9-Aminoanthracene 96.0+%, TCI America™

Catalog No. A1748100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1748100MG 100 mg
A17481G 1 g
A17485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1748100MG Supplier TCI America Supplier No. A1748100MG
Only null left

Chemical Identifiers

CAS 779-03-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00452690
InChI Key LHNICELDCMPPDE-UHFFFAOYSA-N
Synonym 9-Anthramine
PubChem CID 13069
ChEBI CHEBI:50472
IUPAC Name anthracen-9-amine
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2N

Specifications

Melting Point 170°C
Color Green-Yellow
Quantity 100 mg
Formula Weight 193.25
Percent Purity ≥96.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Aminoanthracene
RTECSNumber CB2279000
TSCA No