missing translation for 'onlineSavingsMsg'
Learn More

9-Aminoacridine 97.0+%, TCI America™

Catalog No. p-7118137
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29051G 1 g
A29055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29051G Supplier TCI America Supplier No. A29051G

Chemical Identifiers

CAS 90-45-9
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.24
MDL Number MFCD00037839
InChI Key XJGFWWJLMVZSIG-UHFFFAOYSA-N
Synonym 9-AA
PubChem CID 7019
ChEBI CHEBI:74789
IUPAC Name acridin-9-amine
SMILES NC1=C2C=CC=CC2=NC2=CC=CC=C12

Specifications

Melting Point 238°C
Color Yellow
Quantity 1 g
Formula Weight 194.24
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Aminoacridine
RTECSNumber AR7300000
TSCA No