missing translation for 'onlineSavingsMsg'
Learn More

9-Acridinecarboxylic Acid Hydrate 97.0+%, TCI America™

Catalog No. A15051G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15051G 1 g
A15055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15051G Supplier TCI America Supplier No. A15051G

Chemical Identifiers

CAS 332927-03-4
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.23
MDL Number MFCD00149578
InChI Key IYRYQBAAHMBIFT-UHFFFAOYSA-N
Synonym 9-acridinecarboxylic acid hydrate, acridine-9-carboxylic acid hydrate, acmc-1afs1, c14h9no2.h2o, acridin-9-carboxylic acid hydrate, acridine-9-carboxylic acid hydrate 1:x
PubChem CID 16211687
IUPAC Name acridine-9-carboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=NC2=CC=CC=C12

Specifications

Quantity 1 g
Formula Weight 223.23
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Acridinecarboxylic Acid Hydrate