accessibility menu, dialog, popup

UserName

HOON

CAS RN 332927-03-4

IUPAC Name: acridine-9-carboxylic acid
Synonyms: acridine-9-carboxylic acid
Molecular Weight (g/mol): 223.23
Molecular Formula: C14H9NO2
InChi Key: IYRYQBAAHMBIFT-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)