missing translation for 'onlineSavingsMsg'
Learn More

9-Acetyl-3,6-diiodocarbazole 96.0+%, TCI America™

Catalog No. p-7117699
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20511G 1 g
A20515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20511G Supplier TCI America Supplier No. A20511G

Chemical Identifiers

CAS 606129-89-9
Molecular Formula C14H9I2NO
Molecular Weight (g/mol) 461.04
MDL Number MFCD10000947
InChI Key HAZMJWKYEJRBCO-UHFFFAOYSA-N
PubChem CID 13407350
IUPAC Name 1-(3,6-diiodo-9H-carbazol-9-yl)ethan-1-one
SMILES CC(=O)N1C2=C(C=C(I)C=C2)C2=C1C=CC(I)=C2

Specifications

Melting Point 228°C
Color Yellow
Quantity 1 g
Formula Weight 461.04
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Acetyl-3,6-diiodocarbazole