accessibility menu, dialog, popup

UserName

H3CONII

CAS RN 606129-89-9

IUPAC Name: 1-(3,6-diiodo-9H-carbazol-9-yl)ethan-1-one
Synonyms: 1-(3,6-diiodocarbazol-9-yl)ethanone
Molecular Weight (g/mol): 461.04
Molecular Formula: C14H9I2NO
InChi Key: HAZMJWKYEJRBCO-UHFFFAOYSA-N
SMILES: CC(=O)N1C2=C(C=C(I)C=C2)C2=C1C=CC(I)=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results