missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7-dioxanonanoic Acid 98.0+%, TCI America™
$102.52 - $421.27
Chemical Identifiers
| CAS | 872679-70-4 |
|---|---|
| Molecular Formula | C22H25NO6 |
| Molecular Weight (g/mol) | 399.443 |
| MDL Number | MFCD08064306 |
| InChI Key | QWHLFJJLRVOHTM-UHFFFAOYSA-N |
| Synonym | 9-(Fmoc-amino)-4,7-dioxanonanoic Acid, 3-[2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]propionic Acid, 3-[2-[2-(Fmoc-amino)ethoxy]ethoxy]propionic Acid, Fmoc-NH-PEG2-CH2CH2COOH |
| PubChem CID | 51340973 |
| IUPAC Name | 3-[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)ethoxy]ethoxy]propanoic acid |
| SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCOCCOCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
F0999200MG
|
TCI America
F0999200MG |
200 mg |
Each for $102.52
|
|
|||||
|
F09991G
|
TCI America
F09991G |
1 g |
Each for $421.27
|
|
|||||
Chemical Identifiers
| 872679-70-4 | |
| 399.443 | |
| QWHLFJJLRVOHTM-UHFFFAOYSA-N | |
| 51340973 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCOCCOCCC(=O)O |
| C22H25NO6 | |
| MFCD08064306 | |
| 9-(Fmoc-amino)-4,7-dioxanonanoic Acid, 3-[2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]propionic Acid, 3-[2-[2-(Fmoc-amino)ethoxy]ethoxy]propionic Acid, Fmoc-NH-PEG2-CH2CH2COOH | |
| 3-[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)ethoxy]ethoxy]propanoic acid |
Specifications
| 872679-70-4 | |
| White | |
| MFCD08064306 | |
| 9-(Fmoc-amino)-4,7-dioxanonanoic Acid, 3-[2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]propionic Acid, 3-[2-[2-(Fmoc-amino)ethoxy]ethoxy]propionic Acid, Fmoc-NH-PEG2-CH2CH2COOH | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCOCCOCCC(=O)O | |
| 399.443 | |
| 399.44 | |
| Crystalline Powder |
| 97°C | |
| C22H25NO6 | |
| 200 mg | |
| QWHLFJJLRVOHTM-UHFFFAOYSA-N | |
| 3-[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)ethoxy]ethoxy]propanoic acid | |
| 51340973 | |
| ≥98.0% (HPLC,T) | |
| 9-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7-dioxanonanoic Acid |
Safety and Handling
TSCA : No
Recommended Storage : Freezer