missing translation for 'onlineSavingsMsg'
Learn More

9-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7-dioxanonanoic Acid 98.0+%, TCI America™

Catalog No. F0999200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0999200MG 200 mg
F09991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0999200MG Supplier TCI America Supplier No. F0999200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 872679-70-4
Molecular Formula C22H25NO6
Molecular Weight (g/mol) 399.443
MDL Number MFCD08064306
InChI Key QWHLFJJLRVOHTM-UHFFFAOYSA-N
Synonym 9-(Fmoc-amino)-4,7-dioxanonanoic Acid, 3-[2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]propionic Acid, 3-[2-[2-(Fmoc-amino)ethoxy]ethoxy]propionic Acid, Fmoc-NH-PEG2-CH2CH2COOH
PubChem CID 51340973
IUPAC Name 3-[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)ethoxy]ethoxy]propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCOCCOCCC(=O)O

Specifications

Melting Point 97°C
Color White
Quantity 200 mg
Formula Weight 399.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7-dioxanonanoic Acid
TSCA No
Recommended Storage Freezer