missing translation for 'onlineSavingsMsg'
Learn More

9,9'-Spirobifluorene-2-boronic acid, 98%

Catalog No. AAH6452506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6452506 5 g
AAH6452503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6452506 Supplier Thermo Scientific Chemicals Supplier No. H6452506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 236389-21-2 | C25H17BO2 | 360.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 236389-21-2
Molecular Formula C25H17BO2
Molecular Weight (g/mol) 360.22
MDL Number MFCD13248568
InChI Key WDDLHUWVLROJLA-UHFFFAOYSA-N
Synonym 9,9'-spirobi fluoren-2-ylboronic acid, 9,9'-spirobi 9h-fluorene-2-boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, 9,9'-spirobi fluorene-2-ylboronic acid, boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl, 9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, boronic acid, 9,9'-spirobi 9h-fluoren-2-yl, acmc-209g4k, 9,9'-spirobifluorene-2-boronic acid, 9,9'-spirobi fluorene-2-yl boronic acid
PubChem CID 22564369
IUPAC Name 9,9'-spirobi[fluorene]-2-ylboronic acid
SMILES OB(O)C1=CC2=C(C=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White
Quantity 5 g
Beilstein 10478712
Formula Weight 360.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 9,9'-Spirobifluorene-2-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only